Learn More
Research Products International Corp Tetracycline Hydrochloride, 100 Grams
SDP
Catalog No. 50213676
Encompass_Preferred
Tetracycline Hydrochloride is a broad-spectrum antibiotic. Tetracycline inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit, preventing the attachment of aminoacyl-tRNA to the mRNA-ribosome complex. This interference disrupts the translation process and inhibits bacterial growth. 
Tetracycline HCL is effective against a broad range of bacteria, both Gram-positive and Gram-negative. It is commonly used as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic. It is also used in molecular biology applications, such as the selection of bacteria carrying tetracycline resistance genes in plasmids.
WARNING! This product can expose you to Tetracycline HCl, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-213-676 Supplier Research Products International Corp Supplier No. T17000100.0
Tetracycline Hydrochloride is a broad-spectrum antibiotic. Tetracycline inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit, preventing the attachment of aminoacyl-tRNA to the mRNA-ribosome complex. This interference disrupts the translation process and inhibits bacterial growth. 
Tetracycline HCL is effective against a broad range of bacteria, both Gram-positive and Gram-negative. It is commonly used as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic. It is also used in molecular biology applications, such as the selection of bacteria carrying tetracycline resistance genes in plasmids.
WARNING! This product can expose you to Tetracycline HCl, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.