Learn More
MOLTOX Molecular Toxicology Inc Ampicillin/Tetracycline Master, 20 mL plate, 5 plates/sleeve, Store in Refrigerator
SDP
Catalog No. 50948611
Encompass
For phenotype confirmation of S. typhimurium Ames strains. Minimal Glucose Agar supplemented with histidine, biotin, ampicillin and tetracycline. For use with pAQ1 strains. MOLTOX provides all products necessary to conduct the Salmonella, E. coli, MOLTOX FT™, µAmes and UMU Mutagenicity Assays. All products are formulated and manufactured using the highest quality components and are consistent with the recommendations set forth by the historical literature corresponding to each test type. Certain Ames materials (e.g. STDiscs™ and Lyophilized S9) have been specifically developed in our laboratory for these assays. All products are thoroughly tested for performance in the assay and are accompanied by Quality Control and Formulation statements.
Catalog No. 50-948-611 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 21202
For phenotype confirmation of S. typhimurium Ames strains. Minimal Glucose Agar supplemented with histidine, biotin, ampicillin and tetracycline. For use with pAQ1 strains. MOLTOX provides all products necessary to conduct the Salmonella, E. coli, MOLTOX FT™, µAmes and UMU Mutagenicity Assays. All products are formulated and manufactured using the highest quality components and are consistent with the recommendations set forth by the historical literature corresponding to each test type. Certain Ames materials (e.g. STDiscs™ and Lyophilized S9) have been specifically developed in our laboratory for these assays. All products are thoroughly tested for performance in the assay and are accompanied by Quality Control and Formulation statements.