Learn More
Abcam Tetracyclines ELISA Kit, 96TEST
Supplier:  Abcam AB28526196TEST
Encompass_Preferred
Tetracyclines ELISA Kit is a Competitive ELISA kit for the measurement of Tetracyclines in Tissue, Milk samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-221-5504