Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetradecafluoro-2-methylpentane 99.0+%, TCI America™
SDP

Catalog No. T101225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T101225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T101225ML Supplier TCI America Supplier No. T101225ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 355-04-4
Molecular Formula C6F14
Molecular Weight (g/mol) 338.044
MDL Number MFCD00059905
InChI Key ROVMKEZVKFJNBD-UHFFFAOYSA-N
Synonym Perfluoroisohexane, Perfluoro-2-methylpentane
PubChem CID 67726
IUPAC Name 1,1,1,2,2,3,3,4,5,5,5-undecafluoro-4-(trifluoromethyl)pentane
SMILES C(C(C(C(F)(F)F)(F)F)(F)F)(C(F)(F)F)(C(F)(F)F)F

Specifications

Color Colorless
Boiling Point 58°C
Quantity 25 mL
Formula Weight 338.04
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetradecafluoro-2-methylpentane
EINECSNumber (2)-2366
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.