Learn More
Medchemexpress LLC Tetradecanedioic acid | 821-38-5 | 99.8% | 258.36 | 1 G
SDP
Catalog No. 5000453685
Encompass_Preferred
Tetradecanedioic acid is an endogenous metabolite and belongs to the class of organic compounds known as long-chain fatty acids. It serves as an endogenous biomarker for assessing the activity of organic anion transporting polypeptides (OATPs). It can also act as a candidate biomarker for OATP-mediated active molecule-active molecule interactions and is a key monomer for the synthesis of aliphatic long-chain polyesters.
  • Endogenous metabolite
  • Belongs to the class of long-chain fatty acids
  • Serves as an endogenous biomarker for assessing OATP activity
  • Candidate biomarker for OATP-mediated active molecule-active molecule interactions
  • Key monomer for the synthesis of aliphatic long-chain polyesters
Catalog No. 50-004-53685 Supplier Medchemexpress LLC Supplier No. HYW0118191G
Add to Cart
Add to Cart
Tetradecanedioic acid is an endogenous metabolite and belongs to the class of organic compounds known as long-chain fatty acids. It serves as an endogenous biomarker for assessing the activity of organic anion transporting polypeptides (OATPs). It can also act as a candidate biomarker for OATP-mediated active molecule-active molecule interactions and is a key monomer for the synthesis of aliphatic long-chain polyesters.
  • Endogenous metabolite
  • Belongs to the class of long-chain fatty acids
  • Serves as an endogenous biomarker for assessing OATP activity
  • Candidate biomarker for OATP-mediated active molecule-active molecule interactions
  • Key monomer for the synthesis of aliphatic long-chain polyesters