Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tetradecanoic acid, 2,3-bis[(1-oxododecyl)oxy]propyl ester | 60175-30-6 | MFCD00063570 | 98.0% | 667.05 g/mol | C41H78O6 | 10 MG
SDP

Catalog No. 5000288249
Encompass_Preferred

1,2-Dilauroyl-3-myristoyl-rac-glycerol is a medium-chain triacylglycerol used as a research reagent and an intermediate for synthesizing functional edible oils and related lipid derivatives. It is typically supplied in small, research-scale quantities for laboratory studies.

  • Medium-chain triacylglycerol suitable for lipid synthesis and formulation.
  • High purity (98.0%) suitable for research applications.
  • Chemical formula C41H78O6; molecular weight 667.05 g/mol.
  • Appearance: white to off-white oil.
  • Provided in small research-scale quantities (e.g., 10 mg).

Catalog No. 50-002-88249 Supplier Medchemexpress LLC Supplier No. HY15773810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,2-Dilauroyl-3-myristoyl-rac-glycerol is a medium-chain triacylglycerol used as a research reagent and an intermediate for synthesizing functional edible oils and related lipid derivatives. It is typically supplied in small, research-scale quantities for laboratory studies.

  • Medium-chain triacylglycerol suitable for lipid synthesis and formulation.
  • High purity (98.0%) suitable for research applications.
  • Chemical formula C41H78O6; molecular weight 667.05 g/mol.
  • Appearance: white to off-white oil.
  • Provided in small research-scale quantities (e.g., 10 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.