Learn More
Medchemexpress LLC Tetradecanoic acid, 2,3-dihydroxypropyl ester | 589-68-4 | 250 MG
SDP
Catalog No. 5000421958
Encompass_Preferred
1-Monomyristin acts as an insecticide, enzyme inhibitor, antibacterial, and antifungal agent. It inhibits rat FAAH with an IC50 of 18 μM and rat MAGL with an IC50 of 32 μM. It suppresses the growth of *Staphylococcus aureus*, *Aggregatibacter actinomycetemcomitans*, and *Candida albicans*. This compound is suitable for research related to bacterial infections, fungal infections, renal cancer, prostate adenocarcinoma, and pancreatic cancer.
  • Acts as an insecticide, enzyme inhibitor, antibacterial, and antifungal agent.
  • Inhibits rat FAAH with an IC50 of 18 μM.
  • Inhibits rat MAGL with an IC50 of 32 μM.
  • Suppresses growth of *Staphylococcus aureus*, *Aggregatibacter actinomycetemcomitans*, and *Candida albicans*.
  • Applicable for research in bacterial and fungal infections.
  • Useful in studies related to renal cancer, prostate adenocarcinoma, and pancreatic cancer.
Catalog No. 50-004-21958 Supplier Medchemexpress LLC Supplier No. 250MGHYN2512
1-Monomyristin acts as an insecticide, enzyme inhibitor, antibacterial, and antifungal agent. It inhibits rat FAAH with an IC50 of 18 μM and rat MAGL with an IC50 of 32 μM. It suppresses the growth of *Staphylococcus aureus*, *Aggregatibacter actinomycetemcomitans*, and *Candida albicans*. This compound is suitable for research related to bacterial infections, fungal infections, renal cancer, prostate adenocarcinoma, and pancreatic cancer.
  • Acts as an insecticide, enzyme inhibitor, antibacterial, and antifungal agent.
  • Inhibits rat FAAH with an IC50 of 18 μM.
  • Inhibits rat MAGL with an IC50 of 32 μM.
  • Suppresses growth of *Staphylococcus aureus*, *Aggregatibacter actinomycetemcomitans*, and *Candida albicans*.
  • Applicable for research in bacterial and fungal infections.
  • Useful in studies related to renal cancer, prostate adenocarcinoma, and pancreatic cancer.