Learn More
Apexbio Technology LLC Tetradecanoyl-carnitine-d3-HCl
SDP
Catalog No. 5000570307
Encompass_Preferred
Tetradecanoyl carnitine-d3 HCl (CAS No 1334532-25-0) is a synthetic derivative used in biomedical research primarily involving metabolic studies and lipid metabolism pathways Originating from carnitine metabolism research it targets pathways related to fatty acid transport and oxidation This compound is known for its activity in cell-based assays often in the nanomolar to low micromolar range indicating its potent effects in vitro It is commonly utilized in cell models focusing on metabolic disorders as well as in animal studies for understanding lipid-related mechanisms Additionally it plays a role in drug screening for compounds affecting lipid metabolism Experimental concentrations typically depend on the specific research design allowing flexibility in various experimental frameworks
Catalog No. 50-005-70307 Supplier Apexbio Technology LLC Supplier No. SL10131
Tetradecanoyl carnitine-d3 HCl (CAS No 1334532-25-0) is a synthetic derivative used in biomedical research primarily involving metabolic studies and lipid metabolism pathways Originating from carnitine metabolism research it targets pathways related to fatty acid transport and oxidation This compound is known for its activity in cell-based assays often in the nanomolar to low micromolar range indicating its potent effects in vitro It is commonly utilized in cell models focusing on metabolic disorders as well as in animal studies for understanding lipid-related mechanisms Additionally it plays a role in drug screening for compounds affecting lipid metabolism Experimental concentrations typically depend on the specific research design allowing flexibility in various experimental frameworks