Learn More
Medchemexpress LLC Tetradecyl acetate | 638-59-5 | MFCD00053737 | 256.42 g/mol | C16H32O2 | 50 G
SDP
Catalog No. 5000210664
Encompass_Preferred
Tetradecyl acetate is a naturally occurring insect sex pheromone (myristyl acetate) produced by Ctenopseustis obliquana females and used in pheromone research, mating disruption studies, and as an analytical standard.
  • Sex pheromone produced by Ctenopseustis obliquana females.
  • Used to disrupt mating of pest species.
  • Suitable as an analytical standard for research and analytical applications.
  • Chemical formula C16H32O2; molecular weight 256.42 g/mol.
Catalog No. 50-002-10664 Supplier Medchemexpress LLC Supplier No. HYW04228450G
Tetradecyl acetate is a naturally occurring insect sex pheromone (myristyl acetate) produced by Ctenopseustis obliquana females and used in pheromone research, mating disruption studies, and as an analytical standard.
  • Sex pheromone produced by Ctenopseustis obliquana females.
  • Used to disrupt mating of pest species.
  • Suitable as an analytical standard for research and analytical applications.
  • Chemical formula C16H32O2; molecular weight 256.42 g/mol.