Learn More
Medchemexpress LLC Tetradecyl acetate | 638-59-5 | MFCD00053737 | 98.5% | 256.42 | C16H32O2 | 1 G
SDP
Catalog No. 5000210140
Encompass_Preferred
Tetradecyl acetate is an analytical standard of myristyl acetate used as an insect sex pheromone in research and pheromone-based pest control studies. It is supplied as a high-purity reference standard for analytical and research applications.
  • Analytical standard for research and analysis.
  • Used as insect sex pheromone and for mating disruption studies.
  • High purity suitable for analytical applications.
  • Available in multiple sizes, e.g. 1 G to 100 G.
Catalog No. 50-002-10140 Supplier Medchemexpress LLC Supplier No. HYW0422841G
Tetradecyl acetate is an analytical standard of myristyl acetate used as an insect sex pheromone in research and pheromone-based pest control studies. It is supplied as a high-purity reference standard for analytical and research applications.
  • Analytical standard for research and analysis.
  • Used as insect sex pheromone and for mating disruption studies.
  • High purity suitable for analytical applications.
  • Available in multiple sizes, e.g. 1 G to 100 G.