Learn More
Medchemexpress LLC Tetradecyl acetate | 638-59-5 | MFCD00053737 | 98.5% | 256.42 g/mol | C16H32O2 | 10 G
SDP
Catalog No. 5000210158
Encompass_Preferred
Tetradecyl acetate is an ester (myristyl acetate) used as an analytical standard and as a sex pheromone component. It is used in pheromone research, analytical chemistry, and pest mating disruption studies.
  • Used as an analytical standard for GC/MS and pheromone analysis.
  • High purity promotes reliable analytical results.
  • Available in multiple pack sizes including 10 G for laboratory use.
  • Suitable for research applications involving insect pheromones and mating disruption.
Catalog No. 50-002-10158 Supplier Medchemexpress LLC Supplier No. HYW04228410G
Tetradecyl acetate is an ester (myristyl acetate) used as an analytical standard and as a sex pheromone component. It is used in pheromone research, analytical chemistry, and pest mating disruption studies.
  • Used as an analytical standard for GC/MS and pheromone analysis.
  • High purity promotes reliable analytical results.
  • Available in multiple pack sizes including 10 G for laboratory use.
  • Suitable for research applications involving insect pheromones and mating disruption.