Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tetradecyl acetate | 638-59-5 | MFCD00053737 | 98.5% | 256.42 g/mol | C16H32O2 | 100 G
SDP

Catalog No. 5000210157
Encompass_Preferred

Tetradecyl acetate is a high-purity chemical standard used as a sex pheromone for entomological research, analytical reference, and pest mating-disruption studies. It is supplied with supporting documentation for analytical and safety use.

  • High purity for analytical applications (98.5%).
  • Molecular formula C16H32O2 and molecular weight 256.42 g/mol.
  • Contains CAS number 638-59-5 for chemical identification.
  • Available in multiple pack sizes, including 100 g.
  • Includes datasheet, certificate of analysis, safety data sheet, and spectral reports.
  • Suitable for pheromone research and mating-disruption studies.

Catalog No. 50-002-10157 Supplier Medchemexpress LLC Supplier No. HYW042284100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetradecyl acetate is a high-purity chemical standard used as a sex pheromone for entomological research, analytical reference, and pest mating-disruption studies. It is supplied with supporting documentation for analytical and safety use.

  • High purity for analytical applications (98.5%).
  • Molecular formula C16H32O2 and molecular weight 256.42 g/mol.
  • Contains CAS number 638-59-5 for chemical identification.
  • Available in multiple pack sizes, including 100 g.
  • Includes datasheet, certificate of analysis, safety data sheet, and spectral reports.
  • Suitable for pheromone research and mating-disruption studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.