Learn More
Medchemexpress LLC Tetradecyl acetate (standard) | 638-59-5 | 98.5% | 256.42 | C16H32O2 | 5 G
SDP
Catalog No. 5000210746
Encompass_Preferred
Tetradecyl acetate is an analytical standard and insect sex pheromone used for research and pest-management studies. It is reported as a sex pheromone produced by Ctenopseustis obliquana females and can be used to disrupt mating of pest species.
  • Analytical standard for tetradecyl acetate
  • Used in pheromone and mating disruption studies
  • High purity (98.51%) suitable for analytical applications
  • Available in multiple pack sizes, including 5 g
Catalog No. 50-002-10746 Supplier Medchemexpress LLC Supplier No. HYW0422845G
Tetradecyl acetate is an analytical standard and insect sex pheromone used for research and pest-management studies. It is reported as a sex pheromone produced by Ctenopseustis obliquana females and can be used to disrupt mating of pest species.
  • Analytical standard for tetradecyl acetate
  • Used in pheromone and mating disruption studies
  • High purity (98.51%) suitable for analytical applications
  • Available in multiple pack sizes, including 5 g