Learn More
Apexbio Technology LLC TETRADECYL-B-D-MALTOSIDE-100mg
SDP
Catalog No. 5000897622
Encompass_Preferred
Tetradecyl- -D-maltoside (CAS No 18449-82-6) is a nonionic detergent commonly used in biochemical and biophysical research It originates from the family of alkyl glycosides and functions by solubilizing membrane proteins without denaturing them which is crucial for maintaining their native structure and function This compound is often utilized in studies involving lipid rafts and protein isolation targeting cellular membranes to elucidate signaling pathways Its in vitro activity is typically in the nanomolar to low micromolar range depending on the experimental setup Tetradecyl- -D-maltoside is widely employed in various applications including cell models animal studies and drug screening assays Experimental concentrations typically depend on the specific research design ensuring optimal solubilization while preserving protein functionality
Catalog No. 50-008-97622 Supplier Apexbio Technology LLC Supplier No. C8168100
Tetradecyl- -D-maltoside (CAS No 18449-82-6) is a nonionic detergent commonly used in biochemical and biophysical research It originates from the family of alkyl glycosides and functions by solubilizing membrane proteins without denaturing them which is crucial for maintaining their native structure and function This compound is often utilized in studies involving lipid rafts and protein isolation targeting cellular membranes to elucidate signaling pathways Its in vitro activity is typically in the nanomolar to low micromolar range depending on the experimental setup Tetradecyl- -D-maltoside is widely employed in various applications including cell models animal studies and drug screening assays Experimental concentrations typically depend on the specific research design ensuring optimal solubilization while preserving protein functionality