Learn More
Cayman Chemical Tetradecyl Phosphonate, Purity: ≥98%, CAS: 4671-75-4, 500 mg
SDP
Catalog No. 502890889
Encompass_Preferred
A pan-antagonist of LPA1-3 receptors (IC50 = 10, 5.5, and 3.1 µM, respectively); activates a peroxisome proliferator-activated receptor γ reporter construct 4-fold compared to controls at 10 µM; partially inhibits autotaxin (IC50 = ~3 µM)
Catalog No. 50-289-0889 Supplier Cayman Chemical Supplier No. 10007565500 MG
A pan-antagonist of LPA1-3 receptors (IC50 = 10, 5.5, and 3.1 µM, respectively); activates a peroxisome proliferator-activated receptor γ reporter construct 4-fold compared to controls at 10 µM; partially inhibits autotaxin (IC50 = ~3 µM)