Learn More
Apexbio Technology LLC Tetradecyl Phosphonate 4671-75-4 1g
SDP
Catalog No. 5000091531
Encompass_Preferred
Tetradecyl Phosphonate is a phosphonate-based antagonist targeting lysophosphatidic acid (LPA) receptors LPA1 LPA2 and LPA3 It is designed to inhibit these G protein-coupled receptors thereby blocking LPA-mediated calcium signaling pathways and modulating downstream cellular responses Tetradecyl Phosphonate exerts its biological activity primarily through simultaneous antagonism of LPA receptors Additionally it exhibits partial inhibitory activity against autotaxin and activates the nuclear receptor peroxisome proliferator-activated receptor (PPAR ) Based on these pharmacological properties Tetradecyl Phosphonate serves as a useful tool for research on the biological roles of LPA signaling pathways and their involvement in various disease mechanisms
Catalog No. 50-000-91531 Supplier Apexbio Technology LLC Supplier No. C33431000
Tetradecyl Phosphonate is a phosphonate-based antagonist targeting lysophosphatidic acid (LPA) receptors LPA1 LPA2 and LPA3 It is designed to inhibit these G protein-coupled receptors thereby blocking LPA-mediated calcium signaling pathways and modulating downstream cellular responses Tetradecyl Phosphonate exerts its biological activity primarily through simultaneous antagonism of LPA receptors Additionally it exhibits partial inhibitory activity against autotaxin and activates the nuclear receptor peroxisome proliferator-activated receptor (PPAR ) Based on these pharmacological properties Tetradecyl Phosphonate serves as a useful tool for research on the biological roles of LPA signaling pathways and their involvement in various disease mechanisms