Learn More
Cayman Chemical Tetradecyl Phosphonate, Purity: ≥98%, CAS: 4671-75-4, 1 g
SDP
Catalog No. 502890887
Encompass_Preferred
A pan-antagonist of LPA1-3 receptors (IC50 = 10, 5.5, and 3.1 µM, respectively); activates a peroxisome proliferator-activated receptor γ reporter construct 4-fold compared to controls at 10 µM; partially inhibits autotaxin (IC50 = ~3 µM)
Catalog No. 50-289-0887 Supplier Cayman Chemical Supplier No. 100075651 G
A pan-antagonist of LPA1-3 receptors (IC50 = 10, 5.5, and 3.1 µM, respectively); activates a peroxisome proliferator-activated receptor γ reporter construct 4-fold compared to controls at 10 µM; partially inhibits autotaxin (IC50 = ~3 µM)