Learn More
Medchemexpress LLC Tetradecyl sulfate (sodium) | 1191-50-0 | MFCD00007468 | 98.0% | 317.4 g/mol | C14H29NaO4S | 1 ML
SDP
Catalog No. 5000280801
Encompass_Preferred
Tetradecyl sulfate sodium is an anionic detergent and sclerosant used as a research reagent to study vascular sclerosis and sclerotherapy models, including esophageal varices and varicose veins. It is supplied as a ready-to-use 10 mM solution (1 mL) and in solid mass sizes for laboratory research; not for human or clinical use.
  • Ready-to-use 10 mM solution for convenient dosing
  • Available in solid forms for custom preparation
  • High purity (98.0%) suitable for research applications
  • Used in models of vascular sclerosis and sclerotherapy
  • Accompanied by safety data sheet and handling guidance
Catalog No. 50-002-80801 Supplier Medchemexpress LLC Supplier No. HYW19504810MM/1ML
Tetradecyl sulfate sodium is an anionic detergent and sclerosant used as a research reagent to study vascular sclerosis and sclerotherapy models, including esophageal varices and varicose veins. It is supplied as a ready-to-use 10 mM solution (1 mL) and in solid mass sizes for laboratory research; not for human or clinical use.
  • Ready-to-use 10 mM solution for convenient dosing
  • Available in solid forms for custom preparation
  • High purity (98.0%) suitable for research applications
  • Used in models of vascular sclerosis and sclerotherapy
  • Accompanied by safety data sheet and handling guidance