Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tetradecyl sulfate sodium salt

Catalog No. 2939381G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-393-81G 1 g
29-393-85G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 29-393-81G Supplier Sigma Aldrich Supplier No. 2939381G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Tetradecyl sulfate sodium salt is a detergent-like sclerosant.tetradecyl sulfate sodium salt (sodium tetradecyl sulphate) has been used:
  • To investigate the effects of sclerosant foam preparation and composition on foam structure, the time course of liquid drainage and foam coarsenin
  • In sclerotherapy of telangiectases and reticular veins
  • In situ studies of self-assembled monolayers of adsorbed cationic or anionic surfactant molecules on atomically flat h-terminated si(111) surfaces in aqueous solutions

Specifications

CAS 1191-50-0
Molecular Formula C14H29NaO4S
Linear Formula CH3(CH2)13 OSO3Na
MDL Number MFCD00007468
Quantity 1 g
Synonym Sodium myristyl sulfate; Sodium tetradecyl sulfate
Molecular Weight (g/mol) 316.43
Percent Purity 95%
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.