Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetradecyl Sulfide 98.0+%, TCI America™
SDP

Catalog No. T080525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T080525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T080525G Supplier TCI America Supplier No. T080525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 35599-83-8
Molecular Formula C28H58S
Molecular Weight (g/mol) 426.83
MDL Number MFCD00026608
InChI Key OCTONCPZMJYQLP-UHFFFAOYSA-N
Synonym Ditetradecyl Sulfide, Myristyl Sulfide
PubChem CID 94980
IUPAC Name 1-(tetradecylsulfanyl)tetradecane
SMILES CCCCCCCCCCCCCCSCCCCCCCCCCCCCC

Specifications

Melting Point 54°C
Color White
Quantity 25 g
Formula Weight 426.83
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetradecyl Sulfide
EINECSNumber (2)-0473&(2)-0475
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.