Learn More
Cayman Chemical Tetradecylphosphocholine, Purity: ≥95%, CAS: 77733-28-9, 1 g
SDP
Catalog No. 502936059
Encompass_Preferred
A zwitterionic surfactant; has been used in the solubilization and purification of GPCRs; has been used as the stationary phase in ion chromatography for the determination of inorganic acids; CMC = ~0.12 mM
Catalog No. 50-293-6059 Supplier Cayman Chemical Supplier No. 257001 G
A zwitterionic surfactant; has been used in the solubilization and purification of GPCRs; has been used as the stationary phase in ion chromatography for the determination of inorganic acids; CMC = ~0.12 mM