Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetradecylsuccinic Anhydride 93.0+%, TCI America™
SDP

Catalog No. T009225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T009225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T009225G Supplier TCI America Supplier No. T009225G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 47165-57-1
Molecular Formula C18H32O3
Molecular Weight (g/mol) 296.45
MDL Number MFCD00014549
InChI Key BZECBEKZECEQRI-UHFFFAOYNA-N
PubChem CID 98128
IUPAC Name 3-tetradecyloxolane-2,5-dione
SMILES CCCCCCCCCCCCCCC1CC(=O)OC1=O

Specifications

Melting Point 75°C
Color White-Yellow
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 296.45
Percent Purity ≥93.0% (GC,T)
Chemical Name or Material Tetradecylsuccinic Anhydride
EINECSNumber (2)-0851
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.