Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetradecyltrimethyl-Ammonium Bromide, 98%, MP Biomedicals™

Catalog No. ICN15210980 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15210980 100 g
ICN15210991 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15210980 Supplier MP Biomedicals, Inc Supplier No. 0215210980
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1119-97-7
Molecular Formula C17H38BrN
Molecular Weight (g/mol) 336.40
MDL Number MFCD00011770
InChI Key CXRFDZFCGOPDTD-UHFFFAOYSA-M
Synonym Myristyltrimethylammonium bromide, Trimethyl(tetradecyl)ammonium bromide
PubChem CID 14250
ChEBI CHEBI:3565
SMILES [Br-].CCCCCCCCCCCCCC[N+](C)(C)C

Specifications

Physical Form Powder
Quantity 100 g
Assay Percent Range 98%
IUPAC Name trimethyl(tetradecyl)azanium;bromide
Formula Weight 336.4
Chemical Name or Material Tetradecyltrimethyl-Ammonium Bromide
EINECSNumber 214-291-9
storageNote1 Store Desiccated
Recommended Storage Store at room temperature (15° to 30°C)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.