Learn More
Medchemexpress LLC Tetradecyltrimethylammonium bromide | 1119-97-7 | 98.0% | 100 G
SDP
Catalog No. 5000410003
Encompass_Preferred
Tetradecyltrimethylammonium bromide is an organic building block and a cationic surfactant with an asymmetrical structure. It is used in capillary electrophoresis as a surface active aid for the separation of acid-derived anionic species.
  • Organic building block.
  • Cationic surfactant with an asymmetrical structure.
  • Used in capillary electrophoresis for separating acid-derived anionic species.
  • Requires a minimum concentration of approximately 0.4 mM for optimal results.
  • Facilitates formation of "hemimicelles" on capillary inner wall.
Catalog No. 50-004-10003 Supplier Medchemexpress LLC Supplier No. HYD0839100G
Tetradecyltrimethylammonium bromide is an organic building block and a cationic surfactant with an asymmetrical structure. It is used in capillary electrophoresis as a surface active aid for the separation of acid-derived anionic species.
  • Organic building block.
  • Cationic surfactant with an asymmetrical structure.
  • Used in capillary electrophoresis for separating acid-derived anionic species.
  • Requires a minimum concentration of approximately 0.4 mM for optimal results.
  • Facilitates formation of "hemimicelles" on capillary inner wall.