Learn More
Sigma Organic Chemistry Tetraethylene glycol dimethyl ether | 250G | 143-24-8 | MFCD00008505 | >=99%; >=99.0% (GC)
Supplier: Sigma Organic Chemistry 172405250G

Tetraethylene glycol dimethyl ether, 250G
About This Item:
Synonym(s): 2,5,8,11,14-Pentaoxapentadecane; Bis[2-(2-methoxyethoxy)ethyl] ether; Dimethoxytetraethylene glycol; Dimethyltetraglycol; Tetraglyme
Linear Formula: CH3O(CH2CH2O)4CH3
Molecular Weight: 222.28
CAS Number: 143-24-8
MDL Number: MFCD00008505
UNSPSC Code: 12162002
Purity: >=99%; >=99.0% (GC)
Boiling Point: 275-276 C (lit.)
Melting Point: -30 C (lit.)
Density: 1.009 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.432 (lit.)
Storage: room temp
EINECS Number: 205-594-7
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.