Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraethyl 1,4,8,11-Tetraazacyclotetradecane-1,4,8,11-tetraacetate 97.0+%, TCI America™
SDP

Catalog No. T2540200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2540200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T2540200MG Supplier TCI America Supplier No. T2540200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aza-crown ether reagent for use in chemical research.

Chemical Identifiers

CAS 126320-57-8
Molecular Formula C26H48N4O8
Molecular Weight (g/mol) 544.69
MDL Number MFCD09263316
InChI Key HGPDBLIYOCNCEH-UHFFFAOYSA-N
Synonym 1,4,8,11-tetrakis ethoxycarbonylmethyl-1,4,8,11-tetraazacyclotetradecane, tetaet, tetraethyl 1,4,8,11-tetraazacyclotetradecane-1,4,8,11-tetraacetate, tetraethyl 2,2',2,2'-1,4,8,11-tetraazacyclotetradecane-1,4,8,11-tetrayl tetraacetate, acmc-1c3a0, cyclam-n,n',n',n-tetraacetic acid tetraethyl ester, 1,4,8,11-tetraazacyclotetradecane-1,4,8,11-tetraacetic acid tetraethyl ester, 1,4,8,11-tetrakis ethoxycarbonyl methyl-1,4,8,11-tetraazacyclotetradecane, 1,4,8,11-tetraazacyclotetradecane-n,n',n',n'-tetraacetic acid, tetraethyl ester, min, ethyl 2-4,8,11-tris 2-ethoxy-2-oxoethyl-1,4,8,11-tetraazacyclotetradecan-1-yl acetate
PubChem CID 14434738
IUPAC Name ethyl 2-[4,8,11-tris(2-ethoxy-2-oxoethyl)-1,4,8,11-tetraazacyclotetradecan-1-yl]acetate
SMILES CCOC(=O)CN1CCCN(CC(=O)OCC)CCN(CC(=O)OCC)CCCN(CC(=O)OCC)CC1

Specifications

Melting Point 87°C
Color White-Yellow
Quantity 200 mg
Formula Weight 544.69
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetraethyl 1,4,8,11-Tetraazacyclotetradecane-1,4,8,11-tetraacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.