Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tetraethyl Methylenediphosphonate

Catalog No. 3591815ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-918-15ML 5 mL
35-918-125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 35-918-15ML Supplier Sigma Aldrich Supplier No. 3591815ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for synthesis of: Dual substrate-site inhibitors of 3-deoxy-D-arabino-heptulosonate 7-phosphate synthaseUnnatural alpha-amino acid derivatives containing gem-biphosphonatesBiphenyl sulfonylamino Me bisphosphonic acids as inhibitors of matrix metalloproteinases and bone resorptionLycopene via Wittig-Horner reactionAlkylaminoethylbisphosphinic acids to target farnesyl diphosphate synthaseAromatic bisphosphonates for use as inhibitors of geranylgeranyl diphosphate synthasePrecursor for synthesis of dendritic polyglycerol anions used toward L-selectin inhibition

Specifications

CAS 1660-94-2
Density 1.16 g/mL at 25°C
Boiling Point 171°C to 174°C (11 mmHg, lit.)
Molecular Formula C9H22O6P2
Refractive Index n20/D 1.442
Linear Formula CH2[P(O)(OCH2CH3)2]2
MDL Number MFCD00039887
Quantity 5 mL
Synonym Bis(diethoxyphosphinyl)methane; Methylenediphosphonic acid tetraethylester; NSC 133889
Molecular Weight (g/mol) 288.21 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.