Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tetraethyl orthosilicate

Catalog No. 333859100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-385-9100ML 100 mL
33-385-925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 33-385-9100ML Supplier Sigma Aldrich Supplier No. 333859100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tetraethyl orthosilicate (TEOS) is an inorganic material used as a precursor for the synthesis of silica-based materials by methods such as sol gel technique, due to its high affinity to water. It can also be used in the synthesis of blended membranes.Will interact with dodecylamine in the formation of intercalation compounds of H+-magadiite and used in a study of mixed-metal bioactive glasses.

Specifications

CAS 78-10-4
Density 0.933 g/mL (at 20°C)
Boiling Point 168°C
Molecular Formula C8H20O4Si
Refractive Index n20/D 1.382 (literature)
Linear Formula Si(OC2H5)4
Quantity 100 mL
MDL Number MFCD00009062
Synonym Orthosilicic acid tetraethyl ester; Silicon tetraethoxide; TEOS; Tetraethoxysilane; Tetraethoxysilicon(IV); Tetraethyl silicate; Orthosilicic acid tetraethyl ester; TEOS; Tetraethoxysilane
Molecular Weight (g/mol) 208.33
Percent Purity 99.999% trace metals basis
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.