Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tetraethyl Orthosilicate

Catalog No. 86578250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
86-578-250ML 250 mL
86-578-1L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 86-578-250ML Supplier Sigma Aldrich Supplier No. 86578250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tetraethyl orthosilicate (TEOS) is an inorganic material that can be used as a silica source for the synthesis of silica-based materials such as silicon dioxide, silicon oxycarbides, silanol, siloxane polymer, and organosilicon thin films for a variety of applications. It can also be used in the synthesis of blended membranes, and the production of aerogel. Other applications include coatings for carpets and other objects.Will interact with dodecylamine in the formation of intercalation compounds of H+-magadiite and used in a study of mixed-metal bioactive glasses.

Specifications

CAS 68696-18-4
Molecular Formula C8H20ClN -+ H2O
Refractive Index n20/D 1.382; n20/D 1.383
Linear Formula (C2H5)4N(Cl) -+ H2O
MDL Number MFCD00149992
Quantity 250 mL
Synonym Orthosilicic acid tetraethyl ester; Silicon tetraethoxide; TEOS; Tetraethoxysilane; Tetraethoxysilicon(IV); Tetraethyl silicate; Orthosilicic acid tetraethyl ester; TEOS; Tetraethoxysilane
Molecular Weight (g/mol) 208.33 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.