Learn More
Medchemexpress LLC Tetraethylammonium 50g
SDP
Catalog No. 5000206445
Encompass_Preferred
Tetraethylammonium (iodide) is a quaternary ammonium compound belonging to the class of alkylammonium salts This compound is commonly used as a phase-transfer catalyst in organic synthesis because it facilitates the transfer of reactants between immiscible phases It can also be used as an electrolyte in electrochemical devices and as a surfactant in detergent formulations Its unique chemical properties make it an important ingredient in a variety of industrial applications including pharmaceuticals agrochemicals and materials science
Catalog No. 50-002-06445 Supplier Medchemexpress LLC Supplier No. HYW10125350G
Tetraethylammonium (iodide) is a quaternary ammonium compound belonging to the class of alkylammonium salts This compound is commonly used as a phase-transfer catalyst in organic synthesis because it facilitates the transfer of reactants between immiscible phases It can also be used as an electrolyte in electrochemical devices and as a surfactant in detergent formulations Its unique chemical properties make it an important ingredient in a variety of industrial applications including pharmaceuticals agrochemicals and materials science