Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tetraethylammonium Bicarbonate

Catalog No. 1126825GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-268-25GF 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 11-268-25GF Supplier Sigma Aldrich Supplier No. 1126825GF
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tetraethylammonium bicarbonate is used:In the synthesis of carbamate esters from amines.As a precipitant for synthesizing Cu/ZnO catalysts via coprecipitation method.As an alternative to the traditional phase-transfer system in [18F]radiofluorinations for preventing microreactor blockages.To promote carboxylation of secondary carboxamides bearing a leaving group at the +a-position in the synthesis of oxazolidine-2,4-diones.

Specifications

CAS 5856-62-2
Density 0.944 g/mL at 25°C
Boiling Point 172°C to 174°C
Molecular Formula C4H11NO
Linear Formula C2H5CH(NH2)CH2OH
Quantity 25 g
MDL Number MFCD00064418
Synonym Tetraethylammonium hydrogen carbonate
Molecular Weight (g/mol) 89.14 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.