Learn More
Selleck Chemical LLC Tetraethylammonium bromide
SDP
Catalog No. 5000003387
Encompass_Preferred
Tetraethylammonium bromide (TEAB TEA bromide) is used as the source of tetraethylammonium ions in pharmacological and physiological studies but is also used in organic chemical synthesis It blocks selective potassium channels
Catalog No. 50-000-03387 Supplier Selleck Chemical LLC Supplier No. S628425MG
Tetraethylammonium bromide (TEAB TEA bromide) is used as the source of tetraethylammonium ions in pharmacological and physiological studies but is also used in organic chemical synthesis It blocks selective potassium channels