Learn More
Medchemexpress LLC Tetraethylammonium bromide | 71-91-0 | MFCD00011825 | 98.0% | 210.16 g·mol⁻¹ | C8H20BrN | 100 G
SDP
Catalog No. 5000303913
Encompass_Preferred
Tetraethylammonium bromide (CAS 71-91-0) is a quaternary ammonium salt that appears as a white to off-white solid and is commonly used as a phase-transfer catalyst and biochemical reagent. It has a molecular formula of C8H20BrN, a molecular weight of 210.16 g·mol⁻¹, and is supplied with high purity for laboratory applications.
  • High purity (≥98%) for reliable experimental results
  • White to off-white solid suitable for handling and storage
  • Effective phase-transfer catalyst in organic synthesis
  • Applicable in biochemical and pharmacological research
  • Available in multiple pack sizes to suit lab needs
  • SDS, COA, and data sheet available for safety and quality verification
Catalog No. 50-003-03913 Supplier Medchemexpress LLC Supplier No. HYW014408100G
Tetraethylammonium bromide (CAS 71-91-0) is a quaternary ammonium salt that appears as a white to off-white solid and is commonly used as a phase-transfer catalyst and biochemical reagent. It has a molecular formula of C8H20BrN, a molecular weight of 210.16 g·mol⁻¹, and is supplied with high purity for laboratory applications.
  • High purity (≥98%) for reliable experimental results
  • White to off-white solid suitable for handling and storage
  • Effective phase-transfer catalyst in organic synthesis
  • Applicable in biochemical and pharmacological research
  • Available in multiple pack sizes to suit lab needs
  • SDS, COA, and data sheet available for safety and quality verification