Learn More
Medchemexpress LLC Tetraethylammonium bromide | 71-91-0 | MFCD00011825 | 98.0% | 210.16 g/mol | C8H20BrN | 1000 G
SDP
Catalog No. 5000302980
Encompass_Preferred
Tetraethylammonium bromide is a quaternary ammonium salt used as a laboratory reagent and phase-transfer agent. It supplies the tetraethylammonium cation paired with bromide and is commonly used in synthetic chemistry and ion-channel research. The product is typically supplied as a high-purity crystalline solid for analytical and preparative applications.
  • Acts as a phase-transfer catalyst to facilitate reactions between immiscible phases.
  • Used as a reagent in ion-channel and electrophysiology studies.
  • High purity suitable for analytical and synthetic work.
  • Crystalline solid, readily soluble in polar solvents such as water and alcohols.
  • Available in multiple package sizes for lab-scale and bulk use.
Catalog No. 50-003-02980 Supplier Medchemexpress LLC Supplier No. HYW0144081000G
Tetraethylammonium bromide is a quaternary ammonium salt used as a laboratory reagent and phase-transfer agent. It supplies the tetraethylammonium cation paired with bromide and is commonly used in synthetic chemistry and ion-channel research. The product is typically supplied as a high-purity crystalline solid for analytical and preparative applications.
  • Acts as a phase-transfer catalyst to facilitate reactions between immiscible phases.
  • Used as a reagent in ion-channel and electrophysiology studies.
  • High purity suitable for analytical and synthetic work.
  • Crystalline solid, readily soluble in polar solvents such as water and alcohols.
  • Available in multiple package sizes for lab-scale and bulk use.