Learn More
Medchemexpress LLC Tetraethylammonium bromide | 71-91-0 | MFCD00011825 | 98.0% | 210.16 g/mol | C8H20BrN | 25 G
SDP
Catalog No. 5000303516
Encompass_Preferred
Tetraethylammonium bromide is a quaternary ammonium salt (C8H20BrN) supplied as a white to off-white crystalline solid. It is commonly used as a phase-transfer catalyst and reagent in organic synthesis, as an electrolyte for electrochemical studies, and as a source of tetraethylammonium ions in biochemical and pharmacological research. For research use only.
  • Quaternary ammonium salt useful as a phase-transfer catalyst.
  • Supports organic synthesis and reagent applications.
  • Suitable as an electrolyte in electrochemical experiments.
  • Provides tetraethylammonium ions for biochemical studies.
  • Available in multiple pack sizes, including 25 g.
  • Typical purity 98% for research-grade use.
Catalog No. 50-003-03516 Supplier Medchemexpress LLC Supplier No. 25GHYW014408
Tetraethylammonium bromide is a quaternary ammonium salt (C8H20BrN) supplied as a white to off-white crystalline solid. It is commonly used as a phase-transfer catalyst and reagent in organic synthesis, as an electrolyte for electrochemical studies, and as a source of tetraethylammonium ions in biochemical and pharmacological research. For research use only.
  • Quaternary ammonium salt useful as a phase-transfer catalyst.
  • Supports organic synthesis and reagent applications.
  • Suitable as an electrolyte in electrochemical experiments.
  • Provides tetraethylammonium ions for biochemical studies.
  • Available in multiple pack sizes, including 25 g.
  • Typical purity 98% for research-grade use.