Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tetraethylammonium bromide | 71-91-0 | MFCD00011825 | 98.0% | 210.16 g/mol | C8H20BrN | 250 G
SDP

Catalog No. 5000302342
Encompass_Preferred

Tetraethylammonium bromide is a quaternary ammonium salt supplied for laboratory research as a biochemical reagent. The product is characterized by its defined chemical identity and purity, and is offered in multiple pack sizes for research use.

  • 98.0% purity.
  • CAS 71-91-0 for unambiguous identification.
  • Molecular formula C8H20BrN; molecular weight 210.16 g/mol.
  • SMILES: CC[N+](CC)(CC)CC.[Br-].
  • Available in multiple pack sizes, including 250 g.
  • Intended for laboratory research and reagent applications.

Catalog No. 50-003-02342 Supplier Medchemexpress LLC Supplier No. 250GHYW014408
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetraethylammonium bromide is a quaternary ammonium salt supplied for laboratory research as a biochemical reagent. The product is characterized by its defined chemical identity and purity, and is offered in multiple pack sizes for research use.

  • 98.0% purity.
  • CAS 71-91-0 for unambiguous identification.
  • Molecular formula C8H20BrN; molecular weight 210.16 g/mol.
  • SMILES: CC[N+](CC)(CC)CC.[Br-].
  • Available in multiple pack sizes, including 250 g.
  • Intended for laboratory research and reagent applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.