Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tetraethylammonium bromide | 71-91-0 | MFCD00011825 | 98.0% | 210.16 g·mol⁻¹ | C8H20BrN | 50 G
SDP

Catalog No. 5000302729
Encompass_Preferred

Tetraethylammonium bromide is a quaternary ammonium salt used as a biochemical reagent and research chemical. It is commonly applied as a potassium channel blocker in electrophysiology studies and as a phase-transfer catalyst in organic synthesis.

  • Purity: 98%.
  • Molecular weight: 210.16 g·mol⁻¹.
  • Chemical formula: C8H20BrN.
  • Appearance: white to off-white solid.
  • Typical applications: potassium channel blocker and phase-transfer catalyst.
  • Storage: store at room temperature under nitrogen; in solution, -80°C (2 years) or -20°C (1 year).
  • SDS and datasheet available.

Catalog No. 50-003-02729 Supplier Medchemexpress LLC Supplier No. HYW01440850G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetraethylammonium bromide is a quaternary ammonium salt used as a biochemical reagent and research chemical. It is commonly applied as a potassium channel blocker in electrophysiology studies and as a phase-transfer catalyst in organic synthesis.

  • Purity: 98%.
  • Molecular weight: 210.16 g·mol⁻¹.
  • Chemical formula: C8H20BrN.
  • Appearance: white to off-white solid.
  • Typical applications: potassium channel blocker and phase-transfer catalyst.
  • Storage: store at room temperature under nitrogen; in solution, -80°C (2 years) or -20°C (1 year).
  • SDS and datasheet available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.