Learn More
Tetraethylammonium bromide, 98%
Description
Tetraethylammonium bromide is used as a phase transfer catalyst. It acts as a source of tetraethylammonium ions in pharmacological and physiological studies. It is also employed in organic chemical synthesis. It plays a major role in the preparation of tetraethylammonium superoxide from potassium superoxide for the conversion of primary alkyl halides to dialkyl peroxides. Furthermore, It is utilized as a catalyst in the oxidation of organic sulfide to sulfoxide using o-iodoxybenzoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tetraethylammonium bromide is used as a phase transfer catalyst. It acts as a source of tetraethylammonium ions in pharmacological and physiological studies. It is also employed in organic chemical synthesis. It plays a major role in the preparation of tetraethylammonium superoxide from potassium superoxide for the conversion of primary alkyl halides to dialkyl peroxides. Furthermore, It is utilized as a catalyst in the oxidation of organic sulfide to sulfoxide using o-iodoxybenzoic acid.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents and bases.
Specifications
Specifications
| Melting Point | ∼285°C (decomposition) |
| Density | 1.397 |
| Odor | Mild |
| Quantity | 5000 g |
| Beilstein | 3563430 |
| Sensitivity | Hygroscopic |
| Merck Index | 14,9199 |
| Solubility Information | Soluble in water. |
| Formula Weight | 210.16 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement P261-P280-P305+P351+P338-P304+P340-P405-P501a Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| EINECSNumber | 200-769-4 |
| RTECSNumber | BS5950000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.