Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraethylammonium bromide, 98%

Catalog No. AC161070010
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic Drum
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC161070010 1 kg Plastic bottle
AC161072500 250 g Plastic bottle
AC161070050 5 kg Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC161070010 Supplier Thermo Scientific Chemicals Supplier No. 161070010
  • $223.00
    $195.65 / Each of 1

    Save $27.35

    12% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Phase transfer catalyst | CAS: 71-91-0 | C8H20BrN | 210.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-91-0
Molecular Formula C8H20BrN
Molecular Weight (g/mol) 210.16
MDL Number MFCD00011825
InChI Key HWCKGOZZJDHMNC-UHFFFAOYSA-M
Synonym tetraethylammonium bromide, teab, tea bromide, tetrylammonium bromide, sympatektoman, tetranium, etylon, bromethyl, beparon, etambro
PubChem CID 6285
IUPAC Name tetraethylazanium;bromide
SMILES CC[N+](CC)(CC)CC.[Br-]

Specifications

Melting Point 285.0°C
Assay Percent Range 97.5% min. (Argentometry)
Packaging Plastic bottle
Linear Formula (C2H5)4NBr
Quantity 1 kg
Beilstein 04,104
Fieser 06,568; 13,46
Merck Index 15,9344
Solubility Information Solubility in water: 2795g/L (25°C). Other solubilities: freely soluble in alcohol,chloroform and acetone,slightly soluble in benzene
Formula Weight 210.16
Percent Purity 98%
Chemical Name or Material Tetraethylammonium bromide, 98%
Show More Show Less
Hazard Category HYGROSCOPIC
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-769-4
RTECSNumber BS5950000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.