Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraethylammonium Bromide, BAKER™, J.T. Baker™

Catalog No. 02004416 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-004-416 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-004-416 Supplier Avantor Supplier No. V46807
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

A grade of chemicals for laboratory use

Chemical Identifiers

CAS 71-91-0
Molecular Formula C8H20BrN
Molecular Weight (g/mol) 210.159
InChI Key HWCKGOZZJDHMNC-UHFFFAOYSA-M
Synonym tetraethylammonium bromide, teab, tea bromide, tetrylammonium bromide, sympatektoman, tetranium, etylon, bromethyl, beparon, etambro
PubChem CID 6285
IUPAC Name tetraethylazanium;bromide
SMILES CC[N+](CC)(CC)CC.[Br-]

Specifications

Packaging Poly Bottle
Quantity 500 g
Specific Gravity 1L = 1.40kg
Formula Weight 210.16
Chemical Name or Material Tetraethylammonium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.