Learn More
Selleck Chemical LLC Tetraethylammonium chloride S4489-1g
SDP
Catalog No. 503110209
Encompass_Preferred
Tetraethylammonium (TEA) chloride is a nonspecific potassium channel blocker with antitumor properties
Catalog No. 50-311-0209 Supplier Selleck Chemical LLC Supplier No. S44891G
Tetraethylammonium (TEA) chloride is a nonspecific potassium channel blocker with antitumor properties