Learn More
Apexbio Technology LLC Tetraethylammonium chloride 56-34-8 50mg
SDP
Catalog No. 5000087691
Encompass_Preferred
Tetraethylammonium chloride (CAS 56-34-8) is a quaternary ammonium compound widely used as a potassium channel blocker in pharmacological and physiological studies It inhibits potassium channels by binding to internal and external sites at the channel pore thereby blocking ion conduction This property enables its use in investigations of potassium channel structure function and mutagenesis In vitro tetraethylammonium compounds have been essential for probing channel permeability and gating mechanisms In vivo studies indicate that TEA attenuates taurine-induced vasorelaxation in rat arteries Clinically its ganglionic blocking activity has been explored for circulatory disorders
Catalog No. 50-000-87691 Supplier Apexbio Technology LLC Supplier No. B726250
Tetraethylammonium chloride (CAS 56-34-8) is a quaternary ammonium compound widely used as a potassium channel blocker in pharmacological and physiological studies It inhibits potassium channels by binding to internal and external sites at the channel pore thereby blocking ion conduction This property enables its use in investigations of potassium channel structure function and mutagenesis In vitro tetraethylammonium compounds have been essential for probing channel permeability and gating mechanisms In vivo studies indicate that TEA attenuates taurine-induced vasorelaxation in rat arteries Clinically its ganglionic blocking activity has been explored for circulatory disorders