Learn More
Medchemexpress LLC Tetraethylammonium chloride | 56-34-8 | 98% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB179310MM/1ML
Encompass_Preferred
Tetraethylammonium chloride is a non-selective potassium channel blocker and a good substrate for organic cation transporter (OCTN1). It also exhibits antitumor properties.
  • Inhibits proliferation of C6 and 9L glioma cells.
  • Significantly increases apoptosis.
  • Elevates Bax/Bcl-2 protein ratio.
  • Increases generation of intracellular reactive oxygen species.
  • Significantly increases the amplitude and frequency of contractility of colon and rectum from rats.
Catalog No. 50-004-11719