Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tetraethylammonium chloride | 56-34-8 | 98% | 100 G
SDP

Catalog No. 5000411137
Encompass_Preferred

Tetraethylammonium chloride is a non-selective potassium channel blocker. It is also described as a good substrate for organic cation transporter (OCTN1) and possesses antitumor properties.

  • Molecular weight: 165.70
  • Formula: C8H20ClN
  • Appearance: Solid
  • Color: White to off-white
  • Shipping at room temperature in continental US; may vary elsewhere.
  • Storage at 4°C, sealed, away from moisture. In solvent: -80°C for 6 months, -20°C for 1 month.
  • Soluble in H2O (≥ 100 mg/mL) and DMSO (100 mg/mL).
  • Inhibits proliferation and increases apoptosis in glioma cells.
  • Elevates Bax/Bcl-2 protein ratio and increases intracellular ROS generation in glioma cells.
  • Increases contractility of colon and rectum in rats.

Catalog No. 50-004-11137 Supplier Medchemexpress LLC Supplier No. HYB1793100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetraethylammonium chloride is a non-selective potassium channel blocker. It is also described as a good substrate for organic cation transporter (OCTN1) and possesses antitumor properties.

  • Molecular weight: 165.70
  • Formula: C8H20ClN
  • Appearance: Solid
  • Color: White to off-white
  • Shipping at room temperature in continental US; may vary elsewhere.
  • Storage at 4°C, sealed, away from moisture. In solvent: -80°C for 6 months, -20°C for 1 month.
  • Soluble in H2O (≥ 100 mg/mL) and DMSO (100 mg/mL).
  • Inhibits proliferation and increases apoptosis in glioma cells.
  • Elevates Bax/Bcl-2 protein ratio and increases intracellular ROS generation in glioma cells.
  • Increases contractility of colon and rectum in rats.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.