Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tetraethylammonium chloride | 56-34-8 | 98% | 5 G
SDP

Catalog No. 5000413029
Encompass_Preferred

Tetraethylammonium chloride is a non-selective potassium channel blocker. It also acts as a good substrate for organic cation transporter (OCTN1) and has demonstrated antitumor properties. In vitro studies show it inhibits proliferation and increases apoptosis in glioma cells, while in vivo it affects contractility of rat colon and rectum.

  • Acts as a non-selective potassium channel blocker.
  • Good substrate for organic cation transporter (OCTN1).
  • Exhibits antitumor properties.
  • Inhibits proliferation of C6 and 9L glioma cells.
  • Increases apoptosis in C6 and 9L glioma cells.
  • Elevates Bax/Bcl-2 protein ratio.
  • Increases intracellular ROS in C6 and 9L cells.
  • Increases amplitude and frequency of contractility of rat colon and rectum.

Catalog No. 50-004-13029 Supplier Medchemexpress LLC Supplier No. HYB17935G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetraethylammonium chloride is a non-selective potassium channel blocker. It also acts as a good substrate for organic cation transporter (OCTN1) and has demonstrated antitumor properties. In vitro studies show it inhibits proliferation and increases apoptosis in glioma cells, while in vivo it affects contractility of rat colon and rectum.

  • Acts as a non-selective potassium channel blocker.
  • Good substrate for organic cation transporter (OCTN1).
  • Exhibits antitumor properties.
  • Inhibits proliferation of C6 and 9L glioma cells.
  • Increases apoptosis in C6 and 9L glioma cells.
  • Elevates Bax/Bcl-2 protein ratio.
  • Increases intracellular ROS in C6 and 9L cells.
  • Increases amplitude and frequency of contractility of rat colon and rectum.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.