Learn More
Tetraethylammonium chloride, For electrochemical analysis, ≥99.0%, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| CAS | 56-34-8 |
| Linear Formula | (C2H5)4 N(Cl) |
| Quantity | 5 g |
| Solubility Information | Methanol: 0.1 g/mL, clear, colorless |
| Molecular Weight (g/mol) | 165.70 |
| Formula Weight | 165.70 |
| Percent Purity | ≥99.0% (AT); ≥99.0% |
| Grade | For electrochemical analysis |
| Physical Form | Crystals |
Safety and Handling
| RTECSNumber | BS6125000 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.