Learn More
Abcam Tetraethylammonium chloride (TEA), non-selective K+ channel blocker, 5G
Catalog No. 502707724
Encompass_Preferred
MW 165.7 Da. Non-selective K+ channel blocker (6TM family of K+ channels). Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7724 Supplier Abcam Supplier No. AB1202755G
MW 165.7 Da. Non-selective K+ channel blocker (6TM family of K+ channels). Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement