Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraethylammonium Fluoride Tetrahydrofluoride 97.0+%, TCI America™
SDP

Catalog No. T202710G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T202710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T202710G Supplier TCI America Supplier No. T202710G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 145826-81-9
Molecular Formula C8H24F5N
Molecular Weight (g/mol) 229.279
MDL Number MFCD06797184
InChI Key ANODKKTYHRYRDA-UHFFFAOYSA-M
PubChem CID 11790998
IUPAC Name tetraethylazanium;fluoride;tetrahydrofluoride
SMILES CC[N+](CC)(CC)CC.F.F.F.F.[F-]

Specifications

Color Yellow
Quantity 10 g
UN Number 2922
Formula Weight 229.28
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Tetraethylammonium Fluoride Tetrahydrofluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.