Learn More
Tetraethylammonium hydroxide Solution, ∼1.0M (CH3CH2)4NOH in Water, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| CAS | 77-98-5 |
| Linear Formula | (C2H5)4 N(OH) |
| Quantity | 250 mL |
| MDL Number | MFCD00009024 |
| Synonym | TEA hydroxide |
| Formula Weight | 147.26 |
| Grade | Electrochemical |
| Concentration or Composition (by Analyte or Components) | 1.0 M (CH3 CH2)4 NOH aq. soln. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.