Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tetraethylammonium Hydroxide Solution

Catalog No. 302929500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-292-9500ML 500 mL
30-292-9100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 30-292-9500ML Supplier Sigma Aldrich Supplier No. 302929500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tetraethylammonium hydroxide solution (35 wt. % in H2O) may be used as an organic structure-directing agent (SDA)/template in the synthesis of mesoporous molecular sieves and zeolite samples.

Specifications

CAS 77-98-5
Density 1.023 g/mL at 25°C
Molecular Formula C8H21NO
Linear Formula (C2H5)4N(OH)
Refractive Index n20/D 1.404
Quantity 500 mL
MDL Number MFCD00009024
Synonym TEAOH solution; TEA hydroxide
Molecular Weight (g/mol) 147.26 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.